Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Portable Field Sampler, coating CAR/PDMS
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, red hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (2cm), needle size 23 ga, Autosampler, pk of 1, plain gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, black hub
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 75 μm CAR/PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, black hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPME Fiber Assembly, 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub