Merck
  • Quantitation of anticonvulsant drugs in serum by gas-chromatography on the stationary phase SP-2510.
Quantitation of anticonvulsant drugs in serum by gas-chromatography on the stationary phase SP-2510.
Clinical chemistry (1978-03-01)
W Godolphin, J Thoma
PMID630711
ABSTRACT
A new column packing, SP-2510 DA (Supelco, Inc., Bellefonte, Pa. 16823), is an excellent stationary phase for the determination of a wide variety of anticonvulsant drugs by gas--liquid chromatography without derivatization. However, when uncomplicated extraction procedures are used, serum cholesterol interferes with the determination of primidone. By the simple expedient of adding a short "pre-column" containing another phase (SP-2250 DA) the problem is overcome.