Merck
  • Coordination of early cellular reactions during activation of bone resorption in the rat mandible periosteum: An immunohistochemical study.
Coordination of early cellular reactions during activation of bone resorption in the rat mandible periosteum: An immunohistochemical study.
Heliyon (2017-12-12)
Bassam Hassan, Isabelle Fouilloux, Brigitte Baroukh, Annie Llorens, Martin Biosse Duplan, Marjolaine Gosset, Marc Cherruau, Jean-Louis Saffar
ABSTRACT
The activation step of bone remodeling remains poorly characterized. Activation comprises determination of the site to be remodeled, osteoclast precursor recruitment, their migration to the site of remodeling, and differentiation. These actions involve different compartments and cell types. The aim of this study was to investigate events and cell types involved during activation. We used a bone remodeling model in rats where extractions of the upper jaw molars initiate remodeling of the antagonist lower jaw (mandible) cortex along the periosteum. In this model osteoclastic resorption peaks 4 days after extractions. We previously reported that mast cell activation in the periosteum fibrous compartment is an early event of activation, associated with recruitment of circulating monocyte osteoclast precursors. By using immunohistochemistry, we observed 9 hours after induction a spatially oriented expression of InterCellular Adhesion Molecule-1 in the vessels that was inhibited by antagonists of histamine receptors 1 and 2. It was followed at 12 hours by the recruitment of ED1+ monocytes. In parallel, at 9 hours, Vascular Cellular Adhesion Molecule-1+ fibroblast-like cells scattered in the fibrous compartment of the periosteum between the vessels and the osteogenic compartment increased; these cells may be implicated in osteoclast precursor migration. Receptor Activator of NF KappaB Ligand+ cells increased at 12 hours in the osteogenic compartment and reached a peak at 18 hours. At 24 hours the numbers of osteogenic cells and subjacent osteocytes expressing semaphorin 3a, a repulsive for osteoclast precursors, decreased before returning to baseline at 48 hours. These data show that during activation the two periosteum compartments and several cell types are coordinated to recruit and guide osteoclast precursors towards the bone surface.
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–TRITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Macrophages/Monocytes Antibody, clone ED-1, clone ED-1, Chemicon®, from mouse