Merck
  • Identification of ASB7 as ER stress responsive gene through a genome wide in silico screening for genes with ERSE.
Identification of ASB7 as ER stress responsive gene through a genome wide in silico screening for genes with ERSE.
PloS one (2018-04-10)
Vivek Vishnu Anasa, Madhumathi Manickam, Priti Talwar, Palaniyandi Ravanan
ABSTRACT
The endoplasmic reticulum (ER) not only performs its basic function of regulating calcium homeostasis, lipid biosynthesis, folding, modifying and transporting proteins but also plays a decisive role in regulating multiple cellular processes ranging from cell growth and differentiation to apoptosis and autophagy. Disturbances in ER homeostasis initiate the unfolded protein response (UPR) implicated in the pathogenesis of many human diseases. Drugging the UPR components for therapeutic interventions has received considerable attention. The purpose of this study is to identify genes that are previously unsuspected to be regulated under ER stress. Because ER stress-inducible gene expression is majorly regulated under ERSE elements, we screened human genome by adopting an in silico approach using ERSE elements (I, II, III) as probes and identified 337 candidate genes. Having knowledge of the importance of E3 ubiquitin ligase in the ERAD machinery; we validated our preliminary search by focusing on one of the hits i.e. ASB7 gene that encodes E3 ubiquitin ligase. In HeLa cells, we found that pharmacological induction of ER stress led to an increase in the expression of ASB7 with simultaneous activation of UPR pathways. Although knockdown of ASB7 expression leads to significant reduction in GRP78 and CHOP mRNA levels, it did not protect cells from ER stress-induced cell death. Also, an up-regulation in the expression of pro-inflammatory genes like TNF-α and IL-1β in ASB7 knockdown cells was observed under ER stress. Collectively, our findings suggest that ASB7 is regulated under ER stress and this study also identifies several other genes that could apparently be regulated under ER stress.
Sigma-Aldrich
Tunicamycin from Streptomyces sp.
Sigma-Aldrich
MISSION® esiRNA, targeting human ASB7