  • Search Results
  • Vex-seq: high-throughput identification of the impact of genetic variation on pre-mRNA splicing efficiency.
Vex-seq: high-throughput identification of the impact of genetic variation on pre-mRNA splicing efficiency.
Genome biology (2018-06-03)
Scott I Adamson, Lijun Zhan, Brenton R Graveley
ABSTRACT
Understanding the functional impact of genomic variants is a major goal of modern genetics and personalized medicine. Although many synonymous and non-coding variants act through altering the efficiency of pre-mRNA splicing, it is difficult to predict how these variants impact pre-mRNA splicing. Here, we describe a massively parallel approach we use to test the impact on pre-mRNA splicing of 2059 human genetic variants spanning 110 alternative exons. This method, called variant exon sequencing (Vex-seq), yields data that reinforce known mechanisms of pre-mRNA splicing, identifies variants that impact pre-mRNA splicing, and will be useful for increasing our understanding of genome function.
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes