Merck
  • Meniscus structure in human, sheep, and rabbit for animal models of meniscus repair.
Meniscus structure in human, sheep, and rabbit for animal models of meniscus repair.
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society (2009)
Chevrier A, et al.