Merck
Discovery of S1P agonists with a dihydronaphthalene scaffold.
Bioorganic & medicinal chemistry letters (2011-06-07)
Haruto Kurata, Kensuke Kusumi, Kazuhiro Otsuki, Ryo Suzuki, Masakuni Kurono, Yuka Takada, Hiroki Shioya, Takaki Komiya, Hirotaka Mizuno, Takeji Ono, Hiroshi Hagiya, Masashi Minami, Shinji Nakade, Hiromu Habashita
ABSTRACT
Structure-activity relationship of sphingosine-1-phosphate receptor agonists was examined. Cinnamyl derivative 1 was modified to improve S1P(1) agonistic activity as well as selectivity over S1P(3) agonistic activity. Dihydronaphthalene derivative 10d was identified as a potent S1P(1) receptor agonist with high selectivity against S1P(3) and enhanced efficacy in lowering peripheral lymphocyte counts in mice.
Sigma-Aldrich
Cinnamyl alcohol, 98%
Sigma-Aldrich
Cinnamyl alcohol, ≥98%, FG