Merck
  • Phase I Trials of Anti-ENPP3 Antibody-Drug Conjugates in Advanced Refractory Renal Cell Carcinomas.
Phase I Trials of Anti-ENPP3 Antibody-Drug Conjugates in Advanced Refractory Renal Cell Carcinomas.
Clinical cancer research : an official journal of the American Association for Cancer Research (2018-06-01)
John A Thompson, Robert J Motzer, Ana M Molina, Toni K Choueiri, Elisabeth I Heath, Bruce G Redman, Randeep S Sangha, D Scott Ernst, Roberto Pili, Stella K Kim, Leonard Reyno, Aya Wiseman, Fabio Trave, Banmeet Anand, Karen Morrison, Fernando Doñate, Christian K Kollmannsberger
ABSTRACT
Purpose: To determine the safety, pharmacokinetics, and recommended phase II dose of an antibody-drug conjugate (ADC) targeting ectonucleotide phosphodiesterases-pyrophosphatase 3 (ENPP3) conjugated to monomethyl auristatin F (MMAF) in subjects with advanced metastatic renal cell carcinoma (mRCC).Patients and Methods: Two phase I studies were conducted sequentially with 2 ADCs considered equivalent, hybridoma-derived AGS-16M8F and Chinese hamster ovary-derived AGS-16C3F. AGS-16M8F was administered intravenously every 3 weeks at 5 dose levels ranging from 0.6 to 4.8 mg/kg until unacceptable toxicity or progression. The study was terminated before reaching the MTD. A second study with AGS-16C3F started with the AGS-16M8F bridging dose of 4.8 mg/kg given every 3 weeks.Results: The AGS-16M8F study (n = 26) closed before reaching the MTD. The median duration of treatment was 12 weeks (1.7-83 weeks). One subject had durable partial response (PR; 83 weeks) and 1 subject had prolonged stable disease (48 weeks). In the AGS-16C3F study (n = 34), the protocol-defined MTD was 3.6 mg/kg, but this was not tolerated in multiple doses. Reversible keratopathy was dose limiting and required multiple dose deescalations. The 1.8 mg/kg dose was determined to be safe and was associated with clinically relevant signs of antitumor response. Three of 13 subjects at 1.8 mg/kg had durable PRs (range, 100-143 weeks). Eight subjects at 2.7 mg/kg and 1.8 mg/kg had disease control >37 weeks (37.5-141 weeks).Conclusions: AGS-16C3F was tolerated and had durable antitumor activity at 1.8 mg/kg every 3 weeks. Clin Cancer Res; 24(18); 4399-406. ©2018 AACR.