Merck
  • Low Cell-Matrix Adhesion Reveals Two Subtypes of Human Pluripotent Stem Cells.
Low Cell-Matrix Adhesion Reveals Two Subtypes of Human Pluripotent Stem Cells.
Stem cell reports (2018-07-17)
Leqian Yu, Junjun Li, Jiayin Hong, Yasuhiro Takashima, Nanae Fujimoto, Minako Nakajima, Akihisa Yamamoto, Xiaofeng Dong, Yujiao Dang, Yu Hou, Wei Yang, Itsunari Minami, Keisuke Okita, Motomu Tanaka, Chunxiong Luo, Fuchou Tang, Yong Chen, Chao Tang, Hidetoshi Kotera, Li Liu
ABSTRACT
We show that a human pluripotent stem cell (hPSC) population cultured on a low-adhesion substrate developed two hPSC subtypes with different colony morphologies: flat and domed. Notably, the dome-like cells showed higher active proliferation capacity and increased several pluripotent genes' expression compared with the flat monolayer cells. We further demonstrated that cell-matrix adhesion mediates the interaction between cell morphology and expression of KLF4 and KLF5 through a serum response factor (SRF)-based regulatory double loop. Our results provide a mechanistic view on the coupling among adhesion, stem cell morphology, and pluripotency, shedding light on the critical role of cell-matrix adhesion in the induction and maintenance of hPSC.
Sigma-Aldrich
Anti-MKL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.