Merck
  • Repression of Noxa by Bmi1 contributes to deguelin-induced apoptosis in non-small cell lung cancer cells.
Repression of Noxa by Bmi1 contributes to deguelin-induced apoptosis in non-small cell lung cancer cells.
Journal of cellular and molecular medicine (2018-09-27)
Wei Li, Xinfang Yu, Zhenkun Xia, Xinyou Yu, Li Xie, Xiaolong Ma, Huiling Zhou, Lijun Liu, Jian Wang, Yifeng Yang, Haidan Liu
ABSTRACT
Deguelin, a natural rotenoid isolated from several plants, has been reported to exert anti-tumour effects in various cancers. However, the molecular mechanism of this regulation remains to be fully elucidated. Here, we found that deguelin inhibited the growth of non-small cell lung cancer (NSCLC) cells both in vitro and in vivo by downregulation of Bmi1 expression. Our data showed that Bmi1 is highly expressed in human NSCLC tissues and cell lines. Knockdown of Bmi1 significantly suppressed NSCLC cell proliferation and colony formation. Deguelin treatment attenuated the binding activity of Bmi1 to the Noxa promoter, thus resulting in Noxa transcription and apoptosis activation. Knockdown of Bmi1 promoted Noxa expression and enhanced deguelin-induced apoptosis, whereas overexpression of Bmi1 down-regulated Noxa protein level and deguelin-induced apoptosis. Overall, our study demonstrated a novel apoptotic mechanism for deguelin to exert its anti-tumour activity in NSCLC cells.
Sigma-Aldrich
Protein G Agarose/Salmon Sperm DNA, 2.5 mL, for use in chromatin immunoprecipitations (ChIP assays)
Sigma-Aldrich
Anti-Noxa Mouse mAb (114C307), liquid, clone 114C307, Calbiochem®
Sigma-Aldrich
Normal Rabbit IgG, This Normal Rabbit IgG is validated for use as a negative control in parallel with specific primary antibodies in ELISA, FC, Immunoblotting, IF, IHC, IP.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid