- The amino acid sequence of -crystallin (fraction II) from calf lens.
The amino acid sequence of -crystallin (fraction II) from calf lens.
The Biochemical journal (1972-07-01)
L R Croft
PMID4674126
ABSTRACT
The amino acid sequence of gamma-crystallin (fraction II) from calf lens has been determined; this indicates it to be a single-chain polypeptide of 165 amino acid residues.