Merck
  • Two-dimensional crystallization of histidine-tagged, HIV-1 reverse transcriptase promoted by a novel nickel-chelating lipid.
Two-dimensional crystallization of histidine-tagged, HIV-1 reverse transcriptase promoted by a novel nickel-chelating lipid.
Journal of structural biology (1994-09-01)
E W Kubalek, S F Le Grice, P O Brown
ABSTRACT
Recombinant proteins containing a short stretch of contiguous histidine residues (approximately 6) ("a His-tag") can be specifically bound to N-nitrilotriacetic-acid-chelated nickel ions, providing a convenient general method for their purification. A lipid derivatized with a nickel-chelating head group may provide a general approach to two-dimensional crystallization of the His-tagged proteins, using the lipid layer technique. We have designed a synthetic phospholipid that carries a chelated nickel ion (Ni-NTA-DOPE). His-tagged recombinant HIV-1 reverse transcriptase (HIV-RT) bound specifically to lipid layers containing Ni-NTA-DOPE and formed crystals within minutes from a dilute protein solution. Two-dimensional crystals preserved in negative stain diffracted strongly to approximately 21 A. The projection map computed from averaged Fourier transforms revealed a structure similar in size and shape to a selected projection view of the 3-D structure that was previously determined for HIV-RT by X-ray crystallography.
Product Description
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand
Avanti
18:1 DGS-NTA, 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), chloroform
Avanti
18:1 DGS-NTA, 1,2-dioleoyl-sn-glycero-3-[(N-(5-amino-1-carboxypentyl)iminodiacetic acid)succinyl] (ammonium salt), powder
Avanti
18:1 DGS-NTA(Ni), Avanti Research - A Croda Brand