PL
EN
Applications
Products
Services
Support
Home
Search Results
Capillary gas chromatography determination of benzaldehyde arising from benzyl alcohol used as preservative in injectable formulations.
Capillary gas chromatography determination of benzaldehyde arising from benzyl alcohol used as preservative in injectable formulations.
Journal of Pharmaceutical and Biomedical Analysis (2003)
Kazemifard AG, et al.