Merck
  • Large scale screening and identification of novel Ebolavirus and Marburgvirus entry inhibitors
Large scale screening and identification of novel Ebolavirus and Marburgvirus entry inhibitors
Antimicrobial Agents and Chemotherapy (2016)
Anantpadma M, et al.