Merck
  • Serious anaphylactic reactions due to protamine sulfate: a systematic literature review
Serious anaphylactic reactions due to protamine sulfate: a systematic literature review
Basic and Clinical Pharmacology and Toxicology (2008)
Nybo M and Madsen JS