Merck
  • 5 `RUNX1-3?USP42 chimeric gene in acute myeloid leukemia can occur through an insertion mechanism rather than translocation and may be mediated by genomic segmental duplications
5 `RUNX1-3?USP42 chimeric gene in acute myeloid leukemia can occur through an insertion mechanism rather than translocation and may be mediated by genomic segmental duplications
Molecular Cytogenetics (2014)
Zagaria A, et al.