Merck
  • Intracellular localization and splicing regulation of FUS/TLS are variably affected by amyotrophic lateral sclerosis-linked mutations.
Intracellular localization and splicing regulation of FUS/TLS are variably affected by amyotrophic lateral sclerosis-linked mutations.
Nucleic acids research (2010-11-27)
Yoshihiro Kino, Chika Washizu, Elisa Aquilanti, Misako Okuno, Masaru Kurosawa, Mizuki Yamada, Hiroshi Doi, Nobuyuki Nukina
ABSTRACT
TLS (translocated in liposarcoma), also known as FUS (fused in sarcoma), is an RNA/DNA-binding protein that plays regulatory roles in transcription, pre-mRNA splicing and mRNA transport. Mutations in TLS are responsible for familial amyotrophic lateral sclerosis (ALS) type 6. Furthermore, TLS-containing intracellular inclusions are found in polyglutamine diseases, sporadic ALS, non-SOD1 familial ALS and a subset of frontotemporal lobar degeneration, indicating a pathological significance of TLS in a wide variety of neurodegenerative diseases. Here, we identified TLS domains that determine intracellular localization of the murine TLS. Among them, PY-NLS located in the C-terminus is a strong determinant of intracellular localization as well as splicing regulation of an E1A-derived minigene. Disruption of PY-NLS promoted the formation of cytoplasmic granules that were partially overlapped with stress granules and P-bodies. Some of the ALS-linked mutations altered both intracellular localization and splicing regulation of TLS, while most mutations alone did not affect splicing regulation. However, phospho-mimetic substitution of Ser505 (or Ser513 in human) could enhance the effects of ALS mutations, highlighting interplay between post-translational modification and ALS-linked mutations. These results demonstrate that ALS-linked mutations can variably cause loss of nuclear functions of TLS depending on the degree of impairment in nuclear localization.