Merck
  • Presynaptic localization of neprilysin contributes to efficient clearance of amyloid-beta peptide in mouse brain.
Presynaptic localization of neprilysin contributes to efficient clearance of amyloid-beta peptide in mouse brain.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2004-01-30)
Nobuhisa Iwata, Hiroaki Mizukami, Keiro Shirotani, Yoshie Takaki, Shin-ichi Muramatsu, Bao Lu, Norma P Gerard, Craig Gerard, Keiya Ozawa, Takaomi C Saido
ABSTRACT
A local increase in amyloid-beta peptide (Abeta) is closely associated with synaptic dysfunction in the brain in Alzheimer's disease. Here, we report on the catabolic mechanism of Abeta at the presynaptic sites. Neprilysin, an Abeta-degrading enzyme, expressed by recombinant adeno-associated viral vector-mediated gene transfer, was axonally transported to presynaptic sites through afferent projections of neuronal circuits. This gene transfer abolished the increase in Abeta levels in the hippocampal formations of neprilysin-deficient mice and also reduced the increase in young mutant amyloid precursor protein transgenic mice. In the latter case, Abeta levels in the hippocampal formation contralateral to the vector-injected side were also significantly reduced as a result of transport of neprilysin from the ipsilateral side, and in both sides soluble Abeta was degraded more efficiently than insoluble Abeta. Furthermore, amyloid deposition in aged mutant amyloid precursor protein transgenic mice was remarkably decelerated. Thus, presynaptic neprilysin has been demonstrated to degrade Abeta efficiently and to retard development of amyloid pathology.
Sigma-Aldrich
Anti-β Galactosidase Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-Glutamate receptor 1 Antibody, from rabbit, purified by affinity chromatography