Merck
  • Grass group I pollen allergens (beta-expansins) lack proteinase activity and do not cause wall loosening via proteolysis
Grass group I pollen allergens (beta-expansins) lack proteinase activity and do not cause wall loosening via proteolysis
European Journal of Biochemistry (2001)
Li LC and Cosgrove DJ