Merck
  • Effects of perinatal hypothyroidism on regulation of reelin and brain-derived neurotrophic factor gene expression in rat hippocampus: Role of DNA methylation and histone acetylation.
Effects of perinatal hypothyroidism on regulation of reelin and brain-derived neurotrophic factor gene expression in rat hippocampus: Role of DNA methylation and histone acetylation.
Steroids (2010-07-06)
Li Sui, Bao-Ming Li
ABSTRACT
Thyroid hormones have long been known to play important roles in the development and functions of the central nervous system, however, the precise molecular mechanisms that regulate thyroid hormone-responsive gene expression are not well understood. The present study investigated the role of DNA methylaion and histone acetylation in the effects of perinatal hypothyroidism on regulation of reelin and brain-derived neurotrophic factor (BDNF) gene expression in rat hippocampus. The findings indicated that the activities of DNA methyltransferase (DNMT), methylated reelin and BDNF genes were up-regulated, whereas, the activities of histone acetylases (HAT), the levels of global acetylated histone 3 (H3) and global acetylated histone 4 (H4), and acetylated H3, acetylated H4 at reelin promoter and at BDNF gene promoter for exon II were down-regulated in the hippocampus at the developmental stage of the hypothyroid animals. These results suggest that epigenetic modification of chromatin might underlie the mechanisms of hypothyroidism-induced down-regulation of reelin and BDNF gene expression in developmental rat hippocampus.