Merck
  • Elimination of the adverse effects of urea fertilizer on seed germination, seedling growth, and early plant growth in soil
Elimination of the adverse effects of urea fertilizer on seed germination, seedling growth, and early plant growth in soil
Proceedings of the National Academy of Sciences of the USA (1988)
Bremner JM and Krogmeier MJ