Merck
  • Signal peptide determinants of SecA binding and stimulation of ATPase activity.
Signal peptide determinants of SecA binding and stimulation of ATPase activity.
The Journal of biological chemistry (2000-04-01)
L Wang, A Miller, D A Kendall
ABSTRACT
A signal peptide is required for entry of a preprotein into the secretory pathway, but how it functions in concert with the other transport components is unknown. In Escherichia coli, SecA is a key component of the translocation machinery found in the cytoplasm and at membrane translocation sites. Synthetic signal peptides corresponding to the wild type alkaline phosphatase signal sequence and three sets of model signal sequences varying in hydrophobicity and amino-terminal charge were generated. These were used to establish the requirements for interaction with SecA. Binding to SecA, modulation of SecA conformations sensitive to protease, and stimulation of SecA-lipid ATPase activity occur with functional signal sequences but not with transport-incompetent ones. The extent of SecA interaction is directly related to the hydrophobicity of the signal peptide core region. For signal peptides of moderate hydrophobicity, stimulation of the SecA-lipid ATPase activity is also dependent on amino-terminal charge. The results demonstrate unequivocally that the signal peptide, in the absence of the mature protein, interacts with SecA in aqueous solution and in a lipid bilayer. We show a clear parallel between the hierarchy of signal peptide characteristics that promote interaction with SecA in vitro and the hierarchy of those observed for function in vivo.