Merck
  • A cell migration device that maintains a defined surface with no cellular damage during wound edge generation
A cell migration device that maintains a defined surface with no cellular damage during wound edge generation
Lab on a chip (2009)
Doran MR, et al.