Merck
  • A multicommuted flow system for the determination of dextrose in parenteral and hemodialysis concentrate solutions
A multicommuted flow system for the determination of dextrose in parenteral and hemodialysis concentrate solutions
Journal of Pharmaceutical and Biomedical Analysis (2005)
Knochen M, et al., Knochen M, et al.