Merck
  • FG-4592 Accelerates Cutaneous Wound Healing by Epidermal Stem Cell Activation via HIF-1α Stabilization.
FG-4592 Accelerates Cutaneous Wound Healing by Epidermal Stem Cell Activation via HIF-1α Stabilization.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2018-05-10)
Di Tang, Junhui Zhang, Tiantian Yan, Jingyu Wei, Xupin Jiang, Dongxia Zhang, Qiong Zhang, Jiezhi Jia, Yuesheng Huang
ABSTRACT
Regional hypoxia promptly develops after trauma because of microvascular injury and increased oxygen consumption. This acute hypoxia plays a positive role in early skin wound healing. One of the mechanisms underlying the beneficial effects of acute hypoxia on wound healing may be increased hypoxia-inducible factor-1 (HIF-1α) expression. HIF-1α may affect the wound-healing process through many aspects, including angiogenesis, metabolism, and extra-cellular matrix synthesis and remodelling. Epidermal stem cells (EpSCs) are important participants in wound repair; however, whether these cells are regulated by hypoxia is unclear. This study aimed to elucidate the regulatory mechanism by which hypoxia acts on EpSCs. CCK8 assays, western blots and live cell station observation were employed to compare the viability, proliferation and motility of EpSCs cultured under normoxic conditions (21% O2) with those cultured under hypoxic conditions (2% O2). Moreover, we used FG-4592 (a prolyl hydroxylase inhibitor that stabilizes HIF-1α in normoxia), KC7F2 (a selective inhibitor of HIF-1α transcription) and siRNA against HIF-1α to regulate HIF-1α expression. Acute hypoxia caused EpSCs to switch from a quiescent state to an activated state with higher viability and motility, as well as an earlier proliferation peak. We demonstrated that the HIF-1 signalling pathway mediated hypoxia-induced activation of EpSCs. Finally, the in vivo experiments showed that exogenous FG-4592 effectively accelerates wound healing, shortens healing times and even induces epidermal hyperplasia. This study demonstrated that both hypoxia and exogenous FG-4592 improve EpSC proliferation and motility by stabilizing HIF-1α, and its results suggest that HIF-1α is an important target through which wound healing can be accelerated and that FG-4592 is a promising new drug for wound repair.