Merck
  • Regeneration-associated cell transplantation contributes to tissue recovery in mice with acute ischemic stroke.
Regeneration-associated cell transplantation contributes to tissue recovery in mice with acute ischemic stroke.
PloS one (2019-01-27)
Taira Nakayama, Eiichiro Nagata, Haruchika Masuda, Takayuki Asahara, Shunya Takizawa
ABSTRACT
Various cell-based therapeutic strategies have been investigated for vascular and tissue regeneration after ischemic stroke. We have developed a novel cell population, called regeneration-associated cells (RACs), by quality- and quantity-controlled culture of unfractionated mononuclear cells. RACs were trans-arterially injected into 10-week-old syngeneic male mice at 1, 3, 5 or 7 days after permanent middle cerebral artery occlusion (MCAO) to determine the optimal timing for administration in terms of outcome at day 21. Next, we examined the effects of RACs injection at day 1 after MCAO on neurological deficits, infarct volume, and mediators of vascular regeneration and anti-inflammation at days 7 and 21. Infarct volume at day 21 was significantly reduced by transplantation of RACs at day 1 or 3. RACs injected at day 1 reduced the infarct volume at day 7 and 21. Angiogenesis and anti-inflammatory mediators, VEGF and IL-10, were increased at day 7, and VEGF was still upregulated at day 21. We also observed significantly enhanced ink perfusion in vivo, tube formation in vitro, and definitive endothelial progenitor cell colonies in colony assay. These results suggest that RAC transplantation in MCAO models promoted significant recovery of neural tissues through intensified anti-inflammatory and angiogenic effects.