Merck
  • Search for novel leads for histamine H3-receptor antagonists: oxygen-containing derivatives.
Search for novel leads for histamine H3-receptor antagonists: oxygen-containing derivatives.
Die Pharmazie (1997-07-01)
H Stark, A Hüls, X Ligneau, J M Arrang, J C Schwartz, W Schunack
ABSTRACT
This study was performed in order to develop new leads for antagonists of the histamine H3-receptor subtype. omega-(1 H-Imidazol-4-yl)alkyl derivatives with ester, ketone or alcohol functionality in the side chain were synthesized and tested concerning their H3-receptor antagonist activity on synaptosomes of rat cerebral cortex. The novel compounds, which possess no nitrogen-containing polar group in the side chain of the imidazole moiety, presented moderate to high antagonist potency in vitro. In this series 3-(1 H-imidazol-4-yl)propyl-3-cyclopentylpropanoate (4) was the most potent compound in vitro with -log Ki = 8.5. Unfortunately, no central antagonist H3-receptor activity was detectable for ester derivatives in the in vitro H3-receptor assay based upon measurement of brain N tau-methylhistamine levels after p.o. administration to mice. Some of these novel antagonists are useful tools for investigations on ligand-receptor interaction because of their distinct receptor activities. On the other hand, the ketone derivative 1-(1 H-imidazol-4-yl)-7-phenyl-4-heptanone (9) in vitro presented an ED50-value of 3.5 +/- 1.5 mg/kg p.o. thus proving to be a new lead for further drug investigations. The most potent compounds in vitro and in vivo also showed high H3-receptor selectivity when tested at other histamine receptor subtypes.