Merck
  • Anticancer, antimicrobial and antioxidant potential of sterically tuned bis-N-heterocyclic salts.
Anticancer, antimicrobial and antioxidant potential of sterically tuned bis-N-heterocyclic salts.
Zeitschrift fur Naturforschung. C, Journal of biosciences (2018-12-01)
Noor Ul Huda, Shamsul Islam, Muhammad Zia, Kainaat William, Fakhar I Abbas, Muhammad Ihtisham Umar, Muhammad Adnan Iqbal, Abdul Mannan
ABSTRACT
The current study was conducted to evaluate the antimicrobial, antioxidant, antileishmanial and cytotoxic potential of designed derivatives of 1,1'-(1,3-phenylenebis(methylene))bis(3-alkyl/aryl-1H-benzimidazol-3-ium) salts. The antibacterial potential of the test compounds was investigated against Staphylococcus aureus, Pseudomonas aeruginosa and two methicillin-resistant S. aureus (MRSA) strains (MRSA10, MRSA11), where compound 6 showed the best results. For brine shrimp lethality bioassay (BSLB), compound 6 again showed up to 100% mortality at 200 μg/mL and 56.7% mortality at 6.25 μg/mL. Antileishmanial assay was performed against Leishmania tropica at 20 μg/mL dosage, where 6 showed the most promising activity with 16.26% survival (83.74% mortality; IC50=14.63 μg/mL). The anticancer potential of the selected benzimidazole derivatives was evaluated against two selected cell lines (human colorectal cancer, HCT-116 and breast adenocarcinoma, MCF-7) using sulforhodamine B (SRB) assay. Compound 6 was found to be the most effective cytotoxic compound with 75% inhibition of HCT-116 proliferation at 1 mg/mL concentration. Succinctly, 6 exhibited impressive pharmacological potential that might be attributed to its higher lipophilic character owing to the longer N-substituted alkyl chains when compared to the other test compounds.