Merck
  • Isotope-labeled versus analog internal standard in LC-MS/MS method for tacrolimus determination in human whole blood samples - A compensation of matrix effects.
Isotope-labeled versus analog internal standard in LC-MS/MS method for tacrolimus determination in human whole blood samples - A compensation of matrix effects.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2018-12-12)
Magdalena Bodnar-Broniarczyk, Tomasz Pawiński, Paweł K Kunicki
ABSTRACT
The aim of this work was to develop and to validate LC-MS/MS method for tacrolimus (TAC) determination in whole blood samples using two different types of internal standards (IS): an isotope-labeled TAC13C,D2 and a structural analog ascomycin (ASC). Matrix effects (ME) were evaluated to determine their influence on validation parameters. The LC-MS/MS analyses were performed using a 4000 QTRAP® mass spectrometer (AB Sciex) coupled to a HPLC 1260 Infinity system (Agilent Technologies). The [M + NH4]+ adducts were monitored with mass transitions of: 821.5 → 768.4 m/z for TAC, 809.5 → 756.4 m/z for ASC, and 824.6 → 771.5 m/z for TAC13C,D2. Blood samples were treated with 0.1 mol/L zinc sulfate - acetonitrile (50:50, v/v) then extracted with tert-butyl methyl ether. ME evaluations were performed by preextraction addition (n = 6), postextraction addition (n = 8), and repeated measures (n = 8) of reference solutions, separately for both ISs. ME, absolute recovery (AR) and process efficiency (PE) were calculated. Low (1.5 ng/mL) and high (16 ng/mL) TAC concentrations were tested. The method was successfully calibrated in a range of: 0.5-20 ng/mL. Both ISs provided satisfactory imprecision (<3.09% and <3.63% for TAC13C,D2 and ASC, respectively) and accuracy (99.55-100.63% and 97.35-101.71% for TAC13C,D2 and ASC, respectively). Similar ARs were found for all three compounds yielding: 74.89-76.36% for TAC, 78.37% for TAC13C,D2 and 75.66% for ASC. Significant ME were observed yielding on the average: -16.04% and -29.07% for TAC, -16.64% for TAC13C,D2 and -28.41% for ASC. Consequently, PE was 64.11% and 53.12% for TAC, 65.35% for TAC13C,D2 and 54.18% for ASC. ME for TAC were perfectly compensated (TAC/IS ratio) in each sample resulting in mean percent value of: 0.89% and -0.97% for TAC13C,D2 and ASC, respectively. An IS ascomycin presented a performance equivalent to TAC13C,D2 in LC-MS/MS method developed for TAC monitoring.