Merck
Next-Generation Asymmetric Membranes Using Thin-Film Liftoff.
Nano letters (2019-07-06)
Brian McVerry, Mackenzie Anderson, Na He, Hyukmin Kweon, Chenhao Ji, Shuangmei Xue, Ethan Rao, Chain Lee, Cheng-Wei Lin, Dayong Chen, Dukwoo Jun, Gaurav Sant, Richard B Kaner
ABSTRACT
For the past 30 years, thin-film membrane composites have been the state-of-the-art technology for reverse osmosis, nanofiltration, ultrafiltration, and gas separation. However, traditional membrane casting techniques, such as phase inversion and interfacial polymerization, limit the types of material that are used for the membrane separation layer. Here, we describe a novel thin-film liftoff (T-FLO) technique that enables the fabrication of thin-film composite membranes with new materials for desalination, organic solvent nanofiltration, and gas separation. The active layer is cast separately from the porous support layer, allowing for the tuning of the thickness and chemistry of the active layer. A fiber-reinforced, epoxy-based resin is then cured on top of the active layer to form a covalently bound support layer. Upon submersion in water, the cured membrane lifts off from the substrate to produce a robust, freestanding, asymmetric membrane composite. We demonstrate the fabrication of three novel T-FLO membranes for chlorine-tolerant reverse osmosis, organic solvent nanofiltration, and gas separation. The isolable nature of support and active-layer formation paves the way for the discovery of the transport and selectivity properties of new polymeric materials. This work introduces the foundation for T-FLO membranes and enables exciting new materials to be implemented as the active layers of thin-film membranes, including high-performance polymers, two-dimensional materials, and metal-organic frameworks.