Merck
  • Fc Characteristics Mediate Selective Placental Transfer of IgG in HIV-Infected Women.
Fc Characteristics Mediate Selective Placental Transfer of IgG in HIV-Infected Women.
Cell (2019-06-18)
David R Martinez, Youyi Fong, Shuk Hang Li, Fang Yang, Madeleine F Jennewein, Joshua A Weiner, Erin A Harrell, Jesse F Mangold, Ria Goswami, George R Seage, Galit Alter, Margaret E Ackerman, Xinxia Peng, Genevieve G Fouda, Sallie R Permar
ABSTRACT
The placental transfer of maternal IgG is critical for infant protection against infectious pathogens. However, factors that modulate the placental transfer of IgG remain largely undefined. HIV-infected women have impaired placental IgG transfer, presenting a unique "disruption model" to define factors that modulate placental IgG transfer. We measured the placental transfer efficiency of maternal HIV and pathogen-specific IgG in US and Malawian HIV-infected mothers and their HIV-exposed uninfected and infected infants. We examined the role of maternal HIV disease progression, infant factors, placental Fc receptor expression, IgG subclass, and glycan signatures and their association with placental IgG transfer efficiency. Maternal IgG characteristics, such as binding to placentally expressed Fc receptors FcγRIIa and FcγRIIIa, and Fc region glycan profiles were associated with placental IgG transfer efficiency. Our findings suggest that Fc region characteristics modulate the selective placental transfer of IgG, with implications for maternal vaccine design and infant health.
Sigma-Aldrich
IgG3, Kappa from human myeloma plasma, purified immunoglobulin, >90% (microfluidic capillary gel electrophoresis)
Sigma-Aldrich
IgG2, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)
Sigma-Aldrich
Mouse Anti-Human IgG3, Hinge Specific (HP6047) Biotin Conjugate, liquid, clone HP6047, Calbiochem®
Sigma-Aldrich
Goat Anti-Human IgG Antibody, HRP conjugate, Chemicon®, from goat
Sigma-Aldrich
IgG1, Kappa from human myeloma plasma, purified immunoglobulin, >95% (SDS-PAGE)