Merck
  • Phaseolus vulgaris leucoagglutinin (PHA-L): A neuroanatomical tracer for electron microscopic analysis of synaptic circuitry in the cat's dorsal lateral geniculate nucleus
Phaseolus vulgaris leucoagglutinin (PHA-L): A neuroanatomical tracer for electron microscopic analysis of synaptic circuitry in the cat's dorsal lateral geniculate nucleus
Journal of Electron Microscopy Technique (1990)
Cucchiaro JB and UhlrichDJ