Merck
  • Hordenine: a novel quorum sensing inhibitor and antibiofilm agent against Pseudomonas aeruginosa
Hordenine: a novel quorum sensing inhibitor and antibiofilm agent against Pseudomonas aeruginosa
Journal of Agricultural and Food Chemistry (2018)
Zhou JW, et al.