Merck
  • Co-operative signalling mechanisms required for erythroid precursor expansion in response to erythropoietin and stem cell factor
Co-operative signalling mechanisms required for erythroid precursor expansion in response to erythropoietin and stem cell factor
British Journal of Haematology (2005)
Arcasoy O and Jiang X