Merck
  • Effect of lithium salts on lactate dehydrogenase, adenylate kinase, and 1-phosphofructokinase activities
Effect of lithium salts on lactate dehydrogenase, adenylate kinase, and 1-phosphofructokinase activities
Journal of Enzyme Inhibition and Medicinal Chemistry (2010)
Russell P, et al.