Merck
  • A Mechanism of Adsorption of beta-Nicotinamide Adenine Dinucleotide on Graphene Sheets: Experiment and Theory
A Mechanism of Adsorption of beta-Nicotinamide Adenine Dinucleotide on Graphene Sheets: Experiment and Theory
Chemistry (Weinheim An Der Bergstrasse, Germany) (2009)
Pumera M, et al.