Merck
  • The kinetic characteristics of human and trypanosomatid phosphofructokinases for the reverse reaction
The kinetic characteristics of human and trypanosomatid phosphofructokinases for the reverse reaction
The Biochemical Journal (2019)
Fernandes PM, et al.