Merck
  • Sphingosine-1-Phosphate Receptor 1 Activity Promotes Tumor Growth by Amplifying VEGF-VEGFR2 Angiogenic Signaling.
Sphingosine-1-Phosphate Receptor 1 Activity Promotes Tumor Growth by Amplifying VEGF-VEGFR2 Angiogenic Signaling.
Cell reports (2019-12-12)
Vijay Avin Balaji Ragunathrao, Mumtaz Anwar, Md Zahid Akhter, Alejandra Chavez, De Yu Mao, Viswanathan Natarajan, Sribalaji Lakshmikanthan, Magdalena Chrzanowska-Wodnicka, Arkadiusz Z Dudek, Lena Claesson-Welsh, Jan K Kitajewski, Kishore K Wary, Asrar B Malik, Dolly Mehta
ABSTRACT
The vascular endothelial growth factor-A (VEGF-A)-VEGFR2 pathway drives tumor vascularization by activating proangiogenic signaling in endothelial cells (ECs). Here, we show that EC-sphingosine-1-phosphate receptor 1 (S1PR1) amplifies VEGFR2-mediated angiogenic signaling to enhance tumor growth. We show that cancer cells induce S1PR1 activity in ECs, and thereby, conditional deletion of S1PR1 in ECs (EC-S1pr1-/- mice) impairs tumor vascularization and growth. Mechanistically, we show that S1PR1 engages the heterotrimeric G-protein Gi, which amplifies VEGF-VEGFR2 signaling due to an increase in the activity of the tyrosine kinase c-Abl1. c-Abl1, by phosphorylating VEGFR2 at tyrosine-951, prolongs VEGFR2 retention on the plasmalemma to sustain Rac1 activity and EC migration. Thus, S1PR1 or VEGFR2 antagonists, alone or in combination, reverse the tumor growth in control mice to the level seen in EC-S1pr1-/- mice. Our findings suggest that blocking S1PR1 activity in ECs has the potential to suppress tumor growth by preventing amplification of VEGF-VEGFR2 signaling.
Sigma-Aldrich
SU 5416, ≥98% (HPLC)
Sigma-Aldrich
Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
PF-573228, ≥95% (HPLC)
Sigma-Aldrich
NSC23766 trihydrochloride, ≥97% (HPLC)
Sigma-Aldrich
Imatinib mesylate, ≥98% (HPLC)