Merck
  • FABP5 coordinates lipid signaling that promotes prostate cancer metastasis.
FABP5 coordinates lipid signaling that promotes prostate cancer metastasis.
Scientific reports (2019-12-14)
Gregory Carbonetti, Tessa Wilpshaar, Jessie Kroonen, Keith Studholme, Cynthia Converso, Simon d'Oelsnitz, Martin Kaczocha
ABSTRACT
Prostate cancer (PCa) is defined by dysregulated lipid signaling and is characterized by upregulation of lipid metabolism-related genes including fatty acid binding protein 5 (FABP5), fatty acid synthase (FASN), and monoacylglycerol lipase (MAGL). FASN and MAGL are enzymes that generate cellular fatty acid pools while FABP5 is an intracellular chaperone that delivers fatty acids to nuclear receptors to enhance PCa metastasis. Since FABP5, FASN, and MAGL have been independently implicated in PCa progression, we hypothesized that FABP5 represents a central mechanism linking cytosolic lipid metabolism to pro-metastatic nuclear receptor signaling. Here, we show that the abilities of FASN and MAGL to promote nuclear receptor activation and PCa metastasis are critically dependent upon co-expression of FABP5 in vitro and in vivo. Our findings position FABP5 as a key driver of lipid-mediated metastasis and suggest that disruption of lipid signaling via FABP5 inhibition may constitute a new avenue to treat metastatic PCa.
Product Description
Avanti
12-NBD Stearate, Avanti Research - A Croda Brand, powder