Merck
  • Imaging of normal and pathologic joint synovium using nonlinear optical microscopy as a potential diagnostic tool
Imaging of normal and pathologic joint synovium using nonlinear optical microscopy as a potential diagnostic tool
Journal of Leukocyte Biology (2010)
Tiwari N, et al.