Merck
  • Three-plane description of astroglial populations of OVLT subdivisions in rat: Tanycyte connections to distant parts of third ventricle.
Three-plane description of astroglial populations of OVLT subdivisions in rat: Tanycyte connections to distant parts of third ventricle.
The Journal of comparative neurology (2019-05-03)
Mihály Kálmán, Erzsébet Oszwald, Károly Pócsai
ABSTRACT
This study demonstrates glial and gliovascular markers of organon vasculosum laminae terminalis (OVLT) in three planes. The distribution of glial markers displayed similarities to the subfornical organ. There was an inner part with vimentin- and nestin-immunopositive glia whereas GFAP and the water-channel aquaporin 4 were found at the periphery. This separation indicates different functions of the two regions. The presence of nestin may indicate stem cell-capabilities whereas aquaporin 4 has been reported to promote the osmoreceptor function. Glutamine synthetase immunoreactivity was sparse like in the area postrema and subfornical organ. The laminin and β-dystroglycan immunolabelings altered along the vessels such as in the subfornical organ indicating altering gliovascular relations. The different subdivisions of OVLT received glial processes of different origins. The posterior periventricular zone contained short vimentin-immunopositive processes from the ependyma of the adjacent surface of the third ventricle. The lateral periventricular zone received forceps-like process systems from the anterolateral part of the third ventricle. Most interestingly, the "dorsal cap" received a mixed group of long GFAP- and vimentin-immunopositive processes from a distant part of the third ventricle. The processes may have two functions: a guidance for newly produced cells like radial glia in immature brain and/or a connection between distant parts of the third ventricle and OVLT.
Sigma-Aldrich
Anti-Water Channel Aquaporin 4 antibody produced in rabbit, lyophilized powder, affinity isolated antibody
Sigma-Aldrich
D.E.R. 332, used as embedding medium
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution