Merck
  • Novel mechanistic insights into treadmill exercise based rescue of social defeat-induced anxiety-like behavior and memory impairment in rats.
Novel mechanistic insights into treadmill exercise based rescue of social defeat-induced anxiety-like behavior and memory impairment in rats.
Physiology & behavior (2014-04-16)
Gaurav Patki, Naimesh Solanki, Fatin Atrooz, Amber Ansari, Farida Allam, Brittany Jannise, Jaganmohan Maturi, Samina Salim
ABSTRACT
Social defeat (SD) induced stress causes physiological and behavioral deficits in rodents, including depression and anxiety-like behaviors, as well as memory impairment. Anxiolytic and mood elevating effects of physical exercise are also known. However, rescue effect of physical exercise in social defeat-induced anxiety, depression or memory impairment has not been addressed. The role of epigenetic mechanisms that potentially contribute to these rescue or protective effects is also not known. The present study investigated the effect of moderate treadmill exercise on anxiety-like behavior and memory function in rats subjected to SD using a modified version of the resident-intruder model for social stress (defeat). Changes in histone acetylation and histone-modifying enzymes were examined in hippocampus, amygdala and frontal cortex which are considered critical for anxiety, depression and cognition. Sprague Dawley rats were randomly assigned in four groups; control, exercised, social defeat, social defeat and exercise. At the end of the SD or control exposure lasting 30 min daily for 7 days, one group of SD rats was subjected to treadmill exercise for 2 weeks, whereas the other SD group was handled without exercise. Anxiety-like behavior tests and radial arm water maze test suggested that moderate treadmill exercise rescued social defeat induced anxiety-like behavior and memory impairment. Moreover, exercise normalized SD-induced increase in oxidative stress, most likely by adjusting antioxidant response. Our data suggests involvement of epigenetic mechanisms including histone acetylation of H3 and modulation of methyl-CpG-binding in the hippocampus that might contribute to the rescue effects of exercise in SD-induced behavioral deficits in rats.
Sigma-Aldrich
Anti-Histone H3 Antibody, clone 6.6.2, clone 6.6.2, Upstate®, from mouse
Sigma-Aldrich
Anti-MeCP2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
OxyBlot Protein Oxidation Detection Kit, The OxyBlot Protein Oxidation Detection Kit provides the reagents to perform the immunoblot detection of carbonyl groups introduced into proteins by oxidative reactions with ozone or oxides of nitrogen or by metal catalyzed oxidation.
Sigma-Aldrich
Anti-Mn-SOD Antibody, Upstate®, from rabbit