Merck
  • Influence of ofloxacin, norfloxacin, nalidixic acid, pyromidic acid and pipemidic acid on human gamma-interferon production and blastogenesis.
Influence of ofloxacin, norfloxacin, nalidixic acid, pyromidic acid and pipemidic acid on human gamma-interferon production and blastogenesis.
The Journal of antimicrobial chemotherapy (1986-06-01)
C De Simone, L Baldinelli, M Ferrazzi, S De Santis, L Pugnaloni, F Sorice
ABSTRACT
Several new quinolone derivatives were investigated for their influence on human lymphocyte blastogenesis and gamma-interferon production following concanavalin A stimulation. All the antimicrobials induced inhibition of lymphocyte DNA synthesis. The gamma-interferon measurements showed that nalidixic acid and norfloxacin have a negative influence on lymphokine production and release.