Merck
  • Antibacterial activities of a fosfomycin/tobramycin combination: a novel inhaled antibiotic for bronchiectasis.
Antibacterial activities of a fosfomycin/tobramycin combination: a novel inhaled antibiotic for bronchiectasis.
The Journal of antimicrobial chemotherapy (2009-08-15)
David L MacLeod, Lynn M Barker, Jennifer L Sutherland, Suzanne C Moss, Jesse L Gurgel, Thomas F Kenney, Jane L Burns, William R Baker
ABSTRACT
To compare the in vitro and in vivo activities of a 4:1 (w/w) fosfomycin/tobramycin combination (FTI) with those of fosfomycin and tobramycin alone against cystic fibrosis (CF) and non-CF bronchiectasis pathogens. Clinical isolates of CF Pseudomonas aeruginosa, Staphylococcus aureus, Haemophilus influenzae, Stenotrophomonas maltophilia, Burkholderia cepacia complex, Escherichia coli and Klebsiellia spp. were evaluated by MIC, MBC, post-antibiotic effect (PAE), synergy, time-kill, a rat pneumonia model and spontaneous mutation frequency (SMF). FTI showed high activity against E. coli, H. influenzae, S. aureus and Klebsiella spp. For the S. aureus strains, 75% of which were methicillin resistant (MRSA), FTI had a lower MIC(90) than tobramycin. For P. aeruginosa, FTI had a lower MIC(90) than fosfomycin, but tobramycin was more active than either. Synergy studies showed no antagonism between fosfomycin and tobramycin, and 93% of the isolates demonstrated no interaction. FTI was rapidly bactericidal and exhibited concentration-dependent killing in time-kill studies. In the rat pneumonia model, FTI and tobramycin demonstrated bactericidal killing of P. aeruginosa; both were more active than fosfomycin alone. The SMF for S. aureus resistance to FTI was 2-4 log(10) lower than that for tobramycin and 2-7 log(10) lower than that for fosfomycin. For P. aeruginosa, the FTI SMF was 2-3 log(10) lower than that for fosfomycin and 1-2 log(10) lower than that for tobramycin. FTI is a broad-spectrum antibiotic combination with high activity in vitro and in vivo. These data suggest FTI could be a potential treatment for respiratory infections caused by gram-positive and gram-negative aerobic bacteria.