Merck
  • Polyaniline/multi-walled carbon nanotube composites with core-shell structures as supercapacitor electrode materials
Polyaniline/multi-walled carbon nanotube composites with core-shell structures as supercapacitor electrode materials
Electrochimica Acta (2010)
Zhou Y, et al.