Merck
  • Sonoreactor-based technology for fast high-throughput proteolytic digestion of proteins.
Sonoreactor-based technology for fast high-throughput proteolytic digestion of proteins.
Journal of proteome research (2007-02-03)
R Rial-Otero, R J Carreira, F M Cordeiro, A J Moro, L Fernandes, I Moura, J L Capelo
ABSTRACT
Fast (120 s) and high-throughput (more than six samples at once) in-gel trypsin digestion of proteins using sonoreactor technology has been achieved. Successful protein identification was done by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry, MALDI-TOF-MS. Specific identification of the adenylylsulphate reductase alfa subunit from a complex protein mixture from Desulfovibrio desulfuricans ATCC 27774 was done as a proof of the methodology. The new sample treatment is of easy implementation, saves time and money, and can be adapted to online procedures and robotic platforms.
Product Description
Supelco
ProteoMass Peptide MALDI-MS Calibration Kit, Peptide MALDI MS Calibration Standard