Merck
  • BST2 regulates interferon gamma-dependent decrease in invasion of HTR-8/SVneo cells via STAT1 and AKT signaling pathways and expression of E-cadherin.
BST2 regulates interferon gamma-dependent decrease in invasion of HTR-8/SVneo cells via STAT1 and AKT signaling pathways and expression of E-cadherin.
Cell adhesion & migration (2020-01-21)
Sonam Verma, Amandeep Kaur Kang, Rahul Pal, Satish Kumar Gupta
ABSTRACT
The mechanism by which interferon-gamma (IFN-γ) downregulates trophoblast invasion needs further investigation. Treatment of HTR-8/SVneo cells with IFN-γ led to a decrease in their invasion concomitant with an increased expression of BST2. Silencing of BST2 by siRNA showed a significant increase in their invasion and spreading after treatment with IFN-γ as well as downregulated expression of E-cadherin. Further, STAT1 silencing inhibited the IFN-γ-dependent increase in the expression of BST2 and E-cadherin. Treatment of HTR-8/SVneo cells with IFN-γ led to the activation of AKT, and its inhibition with PI3K inhibitor abrogated IFN-γ-mediated decrease in invasion/spreading and downregulated BST2 and E-cadherin expression. Collectively, IFN-γ decreases the invasion of HTR-8/SVneo cells by STAT1 and AKT activation via increased expression of BST2 and E-cadherin.
Sigma-Aldrich
MISSION® esiRNA, targeting human BST2
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, HEPES Modification, With 4500 mg/L glucose, L-glutamine, and 25 mM HEPES, without sodium bicarbonate and pyruvate, powder, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human STAT1
Roche
cOmplete ULTRA Tablets, Mini, EDTA-free, EASYpack Protease Inhibitor Cocktail, Tablets supplied in foil blister packs.