Merck
  • Knockdown of Lrp1 in RAW264 cells inhibits osteoclast differentiation and osteoclast-osteoblast interactions in vitro.
Knockdown of Lrp1 in RAW264 cells inhibits osteoclast differentiation and osteoclast-osteoblast interactions in vitro.
Biochemical and biophysical research communications (2020-01-23)
Yukihiro Kohara, Ryuma Haraguchi, Riko Kitazawa, Sohei Kitazawa
ABSTRACT
Low density lipoprotein receptor-related protein 1 (LRP1), a multifunctional cell surface protein, is expressed in bone marrow-derived macrophages. While LRP1 is thought to be a suppressor of osteoclast differentiation at late stages, its function at early stages remains unclear. Here we demonstrate that Lrp1 stable knockdown by lentiviral short hairpin RNA in macrophage cell line RAW264 cells inhibited RANKL-induced osteoclast formation and osteoclastic master transcription factor Nfatc1 mRNA expression as assessed by quantitative RT-PCR. Furthermore, knockdown of the Lrp1 gene suppressed not only differentiation, but also proliferation, and inhibitory effects on osteoblastic ALP activity by osteoclast-derived humoral factors. Thus, we propose that LRP1 in macrophages is required for both differentiation into osteoclasts and osteoclast-osteoblast interactions.
Sigma-Aldrich
MISSION® esiRNA, targeting human LRP1
Sigma-Aldrich
MISSION® pLKO.1-puro Non-Mammalian shRNA Control Plasmid DNA, Targets no known mammalian genes