Merck
  • Films made from plasma-modified corn starch: Chemical, mechanical and barrier properties.
Films made from plasma-modified corn starch: Chemical, mechanical and barrier properties.
Carbohydrate polymers (2020-04-04)
Israel Sifuentes-Nieves, Pamela C Flores-Silva, Carlos Gallardo-Vega, Ernesto Hernández-Hernández, Guadalupe Neira-Velázquez, Guadalupe Mendez-Montealvo, Gonzalo Velazquez
ABSTRACT
In this study, the chemical, mechanical and barrier properties of films made from plasma-modified corn starch (MSF) were evaluated as a function of the amylose content (30, 50 and 70 %). SEM analysis revealed the presence of remnant starch granules (RSG) in all films, which promoted the ordering of helices as suggested by the FTIR results. Moreover, XPS analysis was used to identify the oxidation mechanism in all MSF as the atomic proportion of hydroxyl, carbonyl and carboxyl groups changed. Also, the increase of C-C proportions suggested crosslinking in MSF70. TGA analysis indicated low interaction between starch and the plasticizer as the tensile strength and elongation at break diminished in MSF50 and MSF70 due to the low plasticizing effect of glycerol, the oxidation phenomena and the depolymerization of starch chains. However, the crosslinking of MSF70 showed characteristics of rigid films with good hydrophobic performance.
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)