Merck
  • Conversion of fructose into 5-hydroxymethylfurfural (HMF) and its derivatives promoted by inorganic salt in alcohol.
Conversion of fructose into 5-hydroxymethylfurfural (HMF) and its derivatives promoted by inorganic salt in alcohol.
Carbohydrate research (2012-01-24)
Jitian Liu, Yu Tang, Kaigui Wu, Caifeng Bi, Qiu Cui
ABSTRACT
The conversion of D-fructose to 5-hydroxymethylfurfural (HMF) on a 1 mmol scale was achieved in good yield (68%) using NH(4)Cl as catalyst in isopropanol at 120°C. About 3% of 5-i-propoxymethylfurfural was formed. The reaction in ethanol at 100°C on a 10g scale gave a total yield of HMF and 5-ethoxymethylfurfural of 42%. No mineral acid such as H(2)SO(4) and HCl are required.
Sigma-Aldrich
D-(−)-Fructose, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Levulinic acid, 98%
Sigma-Aldrich
Levulinic acid, ≥97%, FG
Sigma-Aldrich
Levulinic acid, natural, 99%, FG